Anti-LY3300054 Recombinant Antibody

Supplier: ProSci Inc.
10-983
PRSI10-983EA 654 EUR
PRSI10-983
Anti-LY3300054 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2-kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR