Anti-POLR3C Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
16-230
PRSI16-230EA 419 EUR
PRSI16-230
Anti-POLR3C Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: POLR3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR