Anti-MS4A2 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
23-650
PRSI23-650EA 419 EUR
PRSI23-650
Anti-MS4A2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MS4A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR