Anti-NFE2L1 Rabbit Polyclonal Antibody
Supplier: ProSci Inc.
31-335
PRSI31-335EA 660 EUR
PRSI31-335
Anti-NFE2L1 Rabbit Polyclonal Antibody
Antibodies
NFE2L1 activates erythroid-specific, globin gene expression. This gene encodes a protein that is involved in globin gene expression in erythrocytes. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene, NFE2L1, and for 'nuclear respiratory factor 1' which has an official symbol of NRF1. Sequence Note: The RefSeq transcript and protein were derived from transcript and genomic sequence to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on alignments.
Antibody isMore Product Information