Anti-IGHG4 Mouse Monoclonal Antibody [clone: IHCG4-1]
Supplier: ProSci Inc.
34-052
PRSI34-052EA 773 EUR
PRSI34-052
Anti-IGHG4 Mouse Monoclonal Antibody [clone: IHCG4-1]
Antibodies
The regions of relatively constant sequence beyond the variable regions of immunoglobulins are termed constant regions (C regions) and are present in both the heavy and light chains. With very few exceptions, the sites of attachment for carbohydrates on immunoglobulins are located in these C regions. These regions also function to hold the variable regions together by using the disulfide bond between them. The C regions facilitate interaction with the antigen by increasing the maximum rotation of the immunoglobulin arms. Reportedly, a large population of patients with recurrent respiratoryMore Product Information