Anti-ISCA2 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
55-216
PRSI55-216EA 620 EUR
PRSI55-216
Anti-ISCA2 Rabbit Polyclonal Antibody
Antibodies
ISCA2 is involved in the assembly of mitochondrial iron-sulfur proteins. Probably involved in the binding of an intermediate of Fe/S cluster assembly (By similarity).

For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: ISCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR