Anti-CPA6 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
55-785
PRSI55-785EA 620 EUR
PRSI55-785
Anti-CPA6 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene belongs to the family of carboxypeptidases, which catalyze the release of C-terminal amino acid, and have functions ranging from digestion of food to selective biosynthesis of neuroendocrine peptides. Polymorphic variants and a reciprocal translocation t(6;8)(q26;q13) involving this gene, have been associated with Duane retraction syndrome.

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: CPA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR