Anti-BEST2 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
63-854
PRSI63-854EA 620 EUR
PRSI63-854
Anti-BEST2 Rabbit Polyclonal Antibody
Antibodies
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:50~100.

Type: Primary
Antigen: BEST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 374-403
Host: Rabbit
Isotype: Ig
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR