Anti-TNF Rabbit Polyclonal Antibody (Biotin)

Supplier: ProSci Inc.
XP-5286BT
PRSIXP-5286BTEA 509 EUR
PRSIXP-5286BT
Anti-TNF Rabbit Polyclonal Antibody (Biotin)
Antibodies
Tumor Necrosis Factor Alpha (TNF alpha) is a protein secreted by lipopolysaccharide stimulated macrophages, and causes tumor necrosis when injected into tumour bearing mice. TNF alpha is believed to mediate pathogenic shock and tissue injury associated with endotoxemia.

Anti-rat TNF-α specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TNF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR