General purpose gloves, A445
Supplier: Portwest
A445RERXL
PRTWA445RERXLEA 55.5 EUR
PRTWA445RERXL
General purpose gloves, A445
Gloves
PVC Gauntlet general handling gloves are fully coated and have a soft, comfortable cotton interlock lining. The coating has a smooth finish and offers excellent abrasion resistance.
  • Available in a choice of three lengths
  • Fully lined with soft and comfortable cotton
  • Abrasion and tear resistant
  • Gauntlet (45 cm) for hand, wrist and arm protection
  • Fully coated for maximum liquid protection
  • Gauge liner of 12 for dexterity
  • ANSI abrasion level 3
Certifications: EN 388:2016 (4121X)