Height Endurance Visor
Supplier: Portwest
PA03CLR
PRTWPA03CLREA 34.5 EUR
PRTWPA03CLR
Height Endurance Visor
Face Shields
Panoramic wrap-around visor for Height Endurance helmets range. It allows the user to wear over prescription glasses.
  • One-piece curved lens with moulded nose bridge
  • Light and robust
  • Anti-scratch coating for added durability
  • Anti-fog coating for additional comfort to the wearer
  • Retail box aids presentation for retail sales
Compatible with Height Endurance Helmets and suitable for wearing over most prescription glasses, this visor features an anti-scratch and anti-fog treatment for added durability.
Certifications: EN 170 (2C-1.2 ).