Mat frames for PurusMat
Supplier: PURUS
PF284701W00 PF384701W00 PF204701W00
PURUPF284701W00EA 432 EUR
PURUPF284701W00 PURUPF384701W00 PURUPF204701W00
Mat frames for PurusMat
Floor Mats Mat Frames
These frames are designed to be used with the Purus mats to create a system that is ideal for any critical area that does not allow direct adhesion to the flooring surface. The frames are constructed of a hard, rigid, PS base material combined with a non skid backing that holds the mats in place on almost any surface.
  • Protect floors from adhesive residue from the purus mats
  • Reduces the risk of slips and falls
  • Non-slip parts made from polymer coated polyester textile
  • Naturally antimicrobial, mould and mildew resistant