Pipette tips, LTS®, for Rainin pipettes
Supplier: RAININ
17001131 17001129 30389271 30389277 30389270 30389200 17001133 17001119 30389215 30389273 30389275 30389278 30389294 30389297 30389299 30389301 30389304 17001128 17000504 17000498 17001118 17000506 17001132 30389211 30389228 30389230 30389234 30389238 30389245 30389255 30389256 30389257 17005862 17007082 17005864 17005867 30389243 30389303
RAIN17001131EA 421 EUR
RAIN17001131 RAIN17001129 RAIN30389271 RAIN30389277 RAIN30389270 RAIN30389200 RAIN17001133 RAIN17001119 RAIN30389215 RAIN30389273 RAIN30389275 RAIN30389278 RAIN30389294 RAIN30389297 RAIN30389299 RAIN30389301 RAIN30389304 RAIN17001128 RAIN17000504 RAIN17000498 RAIN17001118 RAIN17000506 RAIN17001132 RAIN30389211 RAIN30389228 RAIN30389230 RAIN30389234 RAIN30389238 RAIN30389245 RAIN30389255 RAIN30389256 RAIN30389257 RAIN17005862 RAIN17007082 RAIN17005864 RAIN17005867 RAIN30389243 RAIN30389303
Pipette tips, LTS®, for Rainin pipettes
Pipette Tips
These thin-wall tips are designed for use with RAININ Pipet-Lite, Pipet-Plus and EDP3-Plus pipettes with LTS LiteTouch Tip Ejection System. 1200 µl tips are specifically designed for use with RAININ E12-1200 and E8-1200 multi-channel pipettes. The unique design prevents drop formation and eliminates the need for touch-off between dispenses. The tips are available in different packaging options.
  • Virgin PP, sterile or non sterile
  • Wide orifice tips (1,5 mm) minimise shear forces on delicate samples and flow resistance for viscous samples
  • Extended length tips (