Micropacked GC columns
Supplier: Restek
19000 19001 19002 19003 19004 19005 19006 19007 19008 19009 19010 19011 19012 19013 19014 19015 19016 19017 19018 19019 19020 19021 19022 19023 19040 19002-293 19002-900 19003-900 19005-271 19011-853 19013-900 19014-900 19017-291 19019-810
REST19000EA 287 EUR
REST19000 REST19001 REST19002 REST19003 REST19004 REST19005 REST19006 REST19007 REST19008 REST19009 REST19010 REST19011 REST19012 REST19013 REST19014 REST19015 REST19016 REST19017 REST19018 REST19019 REST19020 REST19021 REST19022 REST19023 REST19040 REST19002-293 REST19002-900 REST19003-900 REST19005-271 REST19011-853 REST19013-900 REST19014-900 REST19017-291 REST19019-810
Micropacked GC columns
Chromatography Columns
These micropacked columns are highly efficient and provide good sample capacity. The unsurpassed inertness of SilcoSmooth tubing is based on Siltek deactivation, which allows the column to be flexed and coiled without any fear of chipping or cracking the inert surface.
  • Efficient, inert, and flexible
  • Easy to install due to multiple internal diameters
  • Standard coils fit all instruments
  • No special instrument configurations required
  • Micropacked columns operate at flows exceeding 10 ml/min for trouble-free operation