FID/injector cleaning kit
Supplier: Restek
20120
REST20120EA 85 EUR
REST20120
FID/injector cleaning kit
Chromatography Supplies Chromatography Spare Parts
This FID/injector cleaning kit can be used wih any injection ports.
Delivery information: Kit includes nylon tube brushes of sizes (¹/₈", ³/₁₆" and ¹/₄"), pipe cleaner, stainless steel tube brushes of sizes (³/₈", ³/₁₆" and ¹/₄") and surface brush, stainless steel jet reamers and emery cloth.