Cross-disc inlet seals for Agilent GC's
Supplier: Restek
21009 21010 21011 21012 20474 20475 20476 20477
REST21009EA 87 EUR
REST21009 REST21010 REST21011 REST21012 REST20474 REST20475 REST20476 REST20477
Cross-disc inlet seals for Agilent GC's
Seals Mechanical Seals
Cross disc inlet seals are ideal for high-flow split applications >500 ml/min on Agilent 5890 GCs.
  • Siltek® treatment provides inertness similar to fused silica
The inlet seal at the base of the Agilent 5890/6890/7890 GC injection port contacts the sample and must be changed frequently to prevent adsorption of active compounds. In addition, septum fragments and sample residue accumulate on the disk surface, requiring disk replacement. To reduce breakdown and adsorption of active compounds, Siltek® or gold-plated seals are recommended.