Inlet liners for Agilent GCs
Supplier: Restek
20772-209.1 20774-209.25 20773-209 20914 20912 20972 20973 20802 20706 21156 21157 21700-205 21700 20713-213 20782-211 21032 22401 20726 20823 20824 20979 20995 20996 21022 21023 21033 21110 21111 21701 22400 22402 22403 22404 20706-200.1 20707-200.5 20707-210 20707-211 20707-214 20708-210.25 20709-200.1 20710-200.5 20710-209 20710-210 20710-211 20710-213 20710-214 20710-216 20713-200.5 20713-210 20713-214 20714-214.25 20773-210 20773-211 20773-216 20781-205 20782-205 20782-209 20782-214 20782-216 20783-209.25 20783-211.25 20783-214.25 20801-200.1 20802-210 20802-213 20802-214 20913-214.5 20915-200.5 20973-200.5 20973-202.5 20973-210 20973-213.5 20973-214 20979-213.25 21023-211 21023-213 21033-211 21033-213 21033-214 21110-214.1 21111-210 21111-214 21156-211.10 21156-216.10 21157-209.10 21157-210.10 21157-213 21157-214 21157-216 22401-213 22404-213
REST20772-209.1EA 0 EUR
REST20772-209.1 REST20774-209.25 REST20773-209 REST20914 REST20912 REST20972 REST20973 REST20802 REST20706 REST21156 REST21157 REST21700-205 REST21700 REST20713-213 REST20782-211 REST21032 REST22401 REST20726 REST20823 REST20824 REST20979 REST20995 REST20996 REST21022 REST21023 REST21033 REST21110 REST21111 REST21701 REST22400 REST22402 REST22403 REST22404 REST20706-200.1 REST20707-200.5 REST20707-210 REST20707-211 REST20707-214 REST20708-210.25 REST20709-200.1 REST20710-200.5 REST20710-209 REST20710-210 REST20710-211 REST20710-213 REST20710-214 REST20710-216 REST20713-200.5 REST20713-210 REST20713-214 REST20714-214.25 REST20773-210 REST20773-211 REST20773-216 REST20781-205 REST20782-205 REST20782-209 REST20782-214 REST20782-216 REST20783-209.25 REST20783-211.25 REST20783-214.25 REST20801-200.1 REST20802-210 REST20802-213 REST20802-214 REST20913-214.5 REST20915-200.5 REST20973-200.5 REST20973-202.5 REST20973-210 REST20973-213.5 REST20973-214 REST20979-213.25 REST21023-211 REST21023-213 REST21033-211 REST21033-213 REST21033-214 REST21110-214.1 REST21111-210 REST21111-214 REST21156-211.10 REST21156-216.10 REST21157-209.10 REST21157-210.10 REST21157-213 REST21157-214 REST21157-216 REST22401-213 REST22404-213
Inlet liners for Agilent GCs
Chromatography Supplies Chromatography Inlet Liners
Inlet liners with deactivation improve trace-level analysis that are suitable for agilent GCs equipped with split/splitless inlets. It is made of borosilicate glass.
  • Increase accuracy and precision by preventing loss of sensitive analytes-even when using wool
  • Lower detection limits for a wide range of active compounds is achieved
  • Liner-to-liner reproducibility through consistent manufacturing and extensive testing