GC column-mass spectrometer connector, EZ No-Vent®
Supplier: Restek
21015 21016 21018 21043 21323 22454 22423
REST21015EA 98 EUR
REST21015 REST21016 REST21018 REST21043 REST21323 REST22454 REST22423
GC column-mass spectrometer connector, EZ No-Vent®
Fittings and Connectors
A critical orifice in the EZ No-Vent® connector minimizes the amount of oxygen allowed into the MS source, eliminating the need for purge gas required. This enables to skip the lengthy vent and pump-down cycle.
  • Changes GC-MS columns in minutes without venting
  • Easy to install and maintain
  • No special tools or plumbing required
  • Gold-plated body for inertness
  • High-temperature polyimide ferrules eliminate leaks at the problematic transfer line fitting
Delivery information: Kit includes EZ No-Vent connector, two 0,4 mm ID adaptor ferrules for capillaryMore Product Information