GC columns, chromosorb diatomaceous earth
Supplier: Restek
80415-800 80000-800 80000-810 80128-800 80128-810 80128-820 80128-830 80129-810 80129-820 80129-830 80129-840 80129-845 80129-851 80129-852 80129-870 80401-810 80405-810 80405-840 80409-800 80409-810 80415-810 80415-840 80416-800 80416-810 80416-840 80417-800 80441-800 80441-810 80442-800 80442-810 80442-840 80443-800 80443-810 80445-800 80446-800 80447-800 80448-800 80449-810 80450-800 80477-800 80477-810 80478-800 80478-810 80444-810 80445-810
REST80415-800EA 0 EUR
REST80415-800 REST80000-800 REST80000-810 REST80128-800 REST80128-810 REST80128-820 REST80128-830 REST80129-810 REST80129-820 REST80129-830 REST80129-840 REST80129-845 REST80129-851 REST80129-852 REST80129-870 REST80401-810 REST80405-810 REST80405-840 REST80409-800 REST80409-810 REST80415-810 REST80415-840 REST80416-800 REST80416-810 REST80416-840 REST80417-800 REST80441-800 REST80441-810 REST80442-800 REST80442-810 REST80442-840 REST80443-800 REST80443-810 REST80445-800 REST80446-800 REST80447-800 REST80448-800 REST80449-810 REST80450-800 REST80477-800 REST80477-810 REST80478-800 REST80478-810 REST80444-810 REST80445-810
GC columns, chromosorb diatomaceous earth
Chromatography Columns
These bonded stationary phase packed columns are used in capillary columns which provide a dramatic increase in column quality.
  • Low bleed levels
  • Outer diameter 1/8 inches
  • Longer column lifetimes and short conditioning times
Mesh size is 100/120 Silcoport W, modified version of Chromosorb W with highest inertness and most consistent performance.
To bridge the gap between traditional packed columns and capillary columns, bonded liquid phases for packed columns was developed. Bonded phases are more resistant to oxidation than nonbonded phases becauseMore Product Information