Jaw crushers, BB 100, BB 200 and BB 300
Supplier: Retsch GmbH
20.053.0001 20.053.0002 20.053.0003 20.053.0007 20.053.0009 20.053.0008 20.053.0018 20.054.1001 20.054.1003 20.054.1013 20.053.0043 20.053.0019 20.059.0001
411-2250EA 21540 EUR
411-2250 RETS20.053.0002 RETS20.053.0003 413-0004 RETS20.053.0009 RETS20.053.0008 RETS20.053.0018 412-0198 RETS20.054.1003 RETS20.054.1013 RETS20.053.0043 RETS20.053.0019 RETS20.059.0001
Jaw crushers, BB 100, BB 200 and BB 300
Mills
Robust and powerful forced feed crushers that have been specifically designed for sample preparation of hard and brittle materials in batches or continuously. The crushers are suitable for applications including constructional materials, mineralogy, metallurgy, ceramics and environmental analysis. The crushed sample is collected in a removable stainless steel collector. Safety features include a no-rebound hopper and safety switch and brake to ensure an immediate stop when the unit is opened.
  • High throughput and high degree of size reduction
  • Crushing chamber is accessible via quick