Analytical sieve shakers, AS 200/AS 300
Supplier: Retsch GmbH
30.030.0001 30.033.0001 30.031.0001 30.032.0001
RETS30.030.0001EA 2410 EUR
RETS30.030.0001 RETS30.033.0001 RETS30.031.0001 RETS30.032.0001
Analytical sieve shakers, AS 200/AS 300
Shakers Sieve Shakers
Retsch analytical sieve shakers are used in research and development, quality control of raw materials, interim and finished products, as well as in production monitoring.
  • Sieving with 3-D effect
  • For wet and dry sieving
  • Short sieving times
  • Low noise, maintenance-free
AS 200 basic; favourably priced basic model.
AS 200 digit cA; powerful comfort model with touch-sensitive keypad, digital display and interval sieving.
AS 200 control; as AS 200 digit, but microprocessor controlled, with freely selectable intervals and digital vibration amplitudeMore Product Information