Bottles, narrow neck, square
Supplier: RIXIUS
2-0100-1000-32
RIXI2-0100-1000-32EA 5.7 EUR
RIXI2-0100-1000-32
Bottles, narrow neck, square
Bottles Plastic Bottles
Narrow-neck square bottle made of HDPE.
  • Good chemical resistance
  • Good temperature resistance
  • Robust, natural