Position TrueBlot® magnetic separator, 6
Supplier: Rockland Immunochemicals
TMS-15-50
ROCKTMS-15-50EA 847 EUR
ROCKTMS-15-50
Position TrueBlot® magnetic separator, 6
Beads Magnetic Beads
6x1,5 ml Position magnetic separator
  • Magnetic separation
  • Six 1,5 ml microcentrifuge tubes
A magnetic separation unit designed to accommodate up to six 1,5 ml microcentrifuge tubes in order to separate magnetic particles contained in those tubes. Optimal working volume is up to 1 ml.
Magnetic separator unit can be used to hold, and separate magnetic particles.