Heptafluorobutyric anhydride (HFAA)

Supplier: Roth Carl

Synonyms: HFBA, 2,2,3,3,4,4,4-Heptafluorobutanoic anhydride, Perfluorobutyric anhydride, HFAA

0026.1
ROTH0026.1EA 147 EUR
ROTH0026.1
Heptafluorobutyric anhydride (HFAA)
Heptafluorobutyric anhydride
Formula: C₈F₁₄O₃
Boiling Pt: 108 °C (1013 hPa)
Melting Pt: 14 °C
Density: 1.653 g/cm³ (25 °C)
MDL Number: MFCD00000432
CAS Number: 336-59-4
EINECS: 206-410-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR