Eosin B

Supplier: Roth Carl

Synonyms: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91

0306.1 0306.2 0306.3
ROTH0306.1EA 0 EUR
ROTH0306.1 ROTH0306.2 ROTH0306.3
Eosin B
Eosin B
Formula: C₂₀H₆Br₂N₂Na₂O₉
Melting Pt: > 310 °C
Storage Temperature: Ambient
MDL Number: MFCD00005041
CAS Number: 548-24-3
EINECS: 208-943-1
Merck Index: 12,03638

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR