Alcian blue 8 GS (C.l. 74240)

Supplier: Roth Carl
Warning

3082.1 3082.2 3082.3
ROTH3082.1EA 0 EUR
ROTH3082.1 ROTH3082.2 ROTH3082.3
Alcian blue 8 GS (C.l. 74240)
Alcian blue 8 GX
Formula: C₅₆H₆₈Cl₄CuN₁₆S₄
Storage Temperature: Ambient
MDL Number: MFCD00010720
CAS Number: 33864-99-2
EINECS: 251-705-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR