Ethyl phenylacetate

Supplier: Roth Carl

4434.1
ROTH4434.1EA 0 EUR
ROTH4434.1
Ethyl phenylacetate
Ethyl phenylacetate
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 226
Melting Pt: –29 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009178
CAS Number: 101-97-3
EINECS: 202-993-8
Merck Index: 13,03872

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR