1,6-Diaminohexane

Supplier: Roth Carl
Danger

Synonyms: 1,6-Hexanediamine, Hexamethylenediamine

4465.1
ROTH4465.1EA 0 EUR
ROTH4465.1
1,6-Diaminohexane
1,6-Diaminohexane
Formula: C₆H₁₆N₂
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 39…42 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00008243
CAS Number: 124-09-4
EINECS: 204-679-6
UN: 2280
ADR: 8,III
Merck Index: 13,04713

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR