L(-)-Glutathione (reduced form)

Supplier: Roth Carl

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

6382.1 6382.2
ROTH6382.1EA 0 EUR
ROTH6382.1 ROTH6382.2
L(-)-Glutathione (reduced form)
L(-)-Glutathione (reduced form)
Formula: C₁₀H₁₇N₃O₆S
Melting Pt: 185…195 °C
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR