MOPS-Na

Supplier: Roth Carl

Synonyms: 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt

6927.1 6927.2
ROTH6927.1EA 0 EUR
ROTH6927.1 ROTH6927.2
MOPS-Na
MOPS-Na
Formula: C₇H₁₄NNaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00064350
CAS Number: 71119-22-7
EINECS: 428-420-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR