Osmium tetraoxide 2% in aqueous solution

Supplier: Roth Carl
Danger

Synonyms: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid

7436.1
ROTH7436.1EA 53.5 EUR
ROTH7436.1
Osmium tetraoxide 2% in aqueous solution
Osmium tetraoxide
Formula: O₄Os
Storage Temperature: Refrigerator
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 3287
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR