Graphite

Supplier: Roth Carl

7614.1
ROTH7614.1EA 0 EUR
ROTH7614.1
Graphite
Graphite
Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Density: 2.09…2.23 g/cm³ (20 °C)
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR