Isobutanol

Supplier: Roth Carl
Danger

Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol

9075.1
ROTH9075.1EA 0 EUR
ROTH9075.1
Isobutanol
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 107.9
Melting Pt: –108 °C
Density: 0.802 g/cm³ (20 °C)
Flash Pt: 28
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3 III
Merck Index: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR