Myrcene

Supplier: Roth Carl

Synonyms: 7-Methyl-3-methyleneocta-1,6-diene

9429.1
ROTH9429.1EA 0 EUR
ROTH9429.1
Myrcene
Myrcene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00008908
CAS Number: 123-35-3
EINECS: 204-622-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR