Dispenser Boxes
Supplier: Roth Carl
HY13.1
ROTHHY13.1EA 73.5 EUR
ROTHHY13.1
Dispenser Boxes
Boxes Dispenser Boxes
Dispenser, made of ABS, for PARAFILM® sealing film.
  • Store, dispense, and cut Parafilm®, tape, labels, and Tough-Tags®
  • Designed with a safety razor that slides and cuts with no danger to fingers
  • Dispenser holds one or two 50 mm (2") rolls, or one 100 mm (4") roll