Magnesium chloride

Supplier: Roth Carl

KK36.2
ROTHKK36.2EA 39.1 EUR
ROTHKK36.2
Magnesium chloride
Magnesium chloride
Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412
Melting Pt: 715
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR