Zinc(II) nitrate hexahydrate

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Zinc nitrate hexahydrate

S12313.C4
S12313.C4EA 0 EUR
S12313.C4
Zinc(II) nitrate hexahydrate
Zinc (II) nitrate hexahydrate
Formula: ZnN₂O₆·6H₂O
Boiling Pt: ∼105 °C (1013 hPa)
Melting Pt: 36 °C
Density: 2.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149889
CAS Number: 10196-18-6
EINECS: 231-943-8
UN: 1514
ADR: 5.1,II
Merck Index: 12,10273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR