Transfer pipettes, Samco™ Fine Tip
Supplier: SAMCO
234 231 232-1S 232-20S 232-11 243 231-20S 233
SAMC234EA 0 EUR
SAMC234 SAMC231 SAMC232-1S SAMC232-20S SAMC232-11 SAMC243 SAMC231-20S SAMC233
Transfer pipettes, Samco™ Fine Tip
Pipettes Pasteur/Transfer Pipettes
These pipettes with fine tip and built-in pipette bulb are ideal for precise dispensing of small drops into small-volume containers.
  • Made of shatterproof, non toxic polyethylene
  • Low-affinity surface—reduces loss of cells and valuable proteins due to binding
  • Can be frozen in liquid nitrogen, or heat sealed for storage
  • Ideal for removing cell layers and separating blood samples
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)