Safety blood lancets
Supplier: SARSTEDT
85.1015 85.1018 85.1019 85.1016 85.1017
SARS85.1015EA 0 EUR
SARS85.1015 SARS85.1018 SARS85.1019 SARS85.1016 SARS85.1017
Safety blood lancets
Blood Lancets
Safety blood lancets for safe capillary blood collection and increased patient comfort.
  • User and patient safety
  • Increased patient comfort
  • Designed for targeted blood flow
It reduces the risk of needle injury and cross contamination because the needle point or blade is concealed both before and after use. The sterile device is for single use only and can be safely discarded.
Ordering information: A range of penetration depths and widths are available to accommodate various volume requirements and skin types, ensuring optimum blood flow.