Prefilter attachment
Supplier: Sartorius
16807
SART16807EA 276 EUR
SART16807
Prefilter attachment
Filter Holders
Allows the separation of coarse solids from samples with bacteriological germ detection in a single operation, before and during the actual germ-retaining filtration step.