Sartobind® Lab LC System Adapter Kit
Supplier: Sartorius
SBLAAU01-1
SARTSBLAAU01-1EA 20 EUR
SARTSBLAAU01-1
Sartobind® Lab LC System Adapter Kit
Filters Membrane Filter Capsules
Sartobind® Rapid A Lab incorporates the state-of-the-art in membrane affinity chromatography (AC) technology, offering fast, flexible and future-proof antibody purification. It is an ideal solution for antibody research, as well as downstream process development for novel biologics before starting clinical and commercial production.
  • The fastest matrix. High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes
  • Productivity boost. Maximum dynamic binding capacity supports the preparation of your