Tork W1/W2/W3 Light Duty Cleaning Cloths, Combi-Roll
Supplier: Essity
500137
SCAA500137EA 109 EUR
SCAA500137
Tork W1/W2/W3 Light Duty Cleaning Cloths, Combi-Roll
Wipes Reusable Cleaning Cloths Cloths for General Applications
Multipurpose cleaning cloths for light-duty cleaning tasks. Use these durable cloths wet or dry to clean dust and dirt, and can rinse and reuse them as needed.
  • These disposable cleaning cloths are compatible with most chemical solvents, so no need to change the cleaning products
  • This combi-roll has 600 sheets to avoid runouts and save refill time
Tork light-duty cleaning cloths are an economical, multipurpose wiper. Compatible with most chemical solvents, these durable cloths can be used wet or dry for various light-duty cleaning tasks. They’re softMore Product Information