Precision laboratorium thermometer, stabilisation of the thermometer zero point by molecular-constant, artificial ageing. Measuring range: -100 to +30 °C Division: 1 °C, Length: 305 mm Enclosed type Colourless prismatic capillary form Filing with al 1 * 1 items
Supplier: Ludwig Schneider GmbH & CO. KG