Thermometer precision laboratory, accu-safe, enclosed scale cap: colourless prismatic ø8-10mm, DE made, with works-calib. cert. with sr. no. and cert. of conformity, packed in square plastic sleeve, with 3 calib. pts, -5to+100 °c, 0.2 °c, 420mm 1 * 1 items
Supplier: Ludwig Schneider GmbH & CO. KG