Thermometer precision lab, accu-safe, with safety-coating, stemform, cap: white back round ø6-8mm, highly transparent, stirrable, f or calib. with sr. no. and cert. of conformity, packed in square plastic sleeve, -10to+100 °c, 0.5 °c, 290mm 1 * 1 items
Supplier: Ludwig Schneider GmbH & CO. KG