DURAN® Blanks for Distillation Flasks, Engler Type
Supplier: DWK Life Sciences
216512805
SCOT216512805EA 2230 EUR
SCOT216512805
DURAN® Blanks for Distillation Flasks, Engler Type
Flasks Distillation Flasks
DURAN® borosilicate glass 3.3, clear.
  • With beaded rim
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.