FLEX starter sets, SymLine®
Supplier: S.C.A.T.
106690 106691 106694
SCTV106690EA 125 EUR
SCTV106690 SCTV106691 SCTV106694
FLEX starter sets, SymLine®
Safety Cans
The flexible waste disposal system for HPLC workstations. Requiring only a few basic elements, this modular system can be adapted to suit virtually any workplace.
  • Simple installation, with table or wall mounting possible
  • Do-It-Yourself modular system is designed for its suitability for any workplace
  • A minimum of components, with many applications
  • Can be flexibly integrated at any time; react quickly to any changes in the laboratory
  • Whether adding additional workplaces or a reinstallation at another location, the system can be made to fit
  • Anti-static and