Syringes with Luer fittings, PP, medi syst
Supplier: SEMADENI
0900
SEMA0900EA 27.9 EUR
SEMA0900
Syringes with Luer fittings, PP, medi syst
Syringes Syringes, Medical
Syringes with three finger rings for a comfortable, precise and residue-free dosage.
  • Graduated scale engraved
  • Easy to disassemble and clean
  • Perfect sealing and easy piston stroke
  • Equipped with a Luer fitting and conical tip
  • With lid lock preventing the piston from jumping
Low pressure is sufficient to inject or dose products, even viscous.