Staining tray
Supplier: SEMADENI
4332
SEMA4332EA 80.2 EUR
SEMA4332
Staining tray
Staining Boxes
Transparent base with a well fitting PP lid.
  • Usable up to 70 °C
  • Stackable with two handles
  • Good chemical resistance
Suitable for dying, bleaching and fixing of gels and membranes susceptible electrophoresis.