Stomacher 400 EVO, laboratory blender for food microbiologists. Unique one-touch bag loading system for quiet and efficient blending. Features easy clean interior with removable paddles, drip tray and access to paddle chamber. Designed for use with b 1 * 1 items
Supplier: Seward (FermionX)